4-Iodo-3-(trifluoromethoxy)benzoic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Carboxy-2-iodo-α,α,α-trifluoroanisole
PC9914-1G PC9914-5G
APOSPC9914-1GEA 910 SEK
APOSPC9914-1G APOSPC9914-5G
4-Iodo-3-(trifluoromethoxy)benzoic acid
4-Iodo-3-(trifluoromethoxy)benzoic acid
Formel: C₈H₄F₃IO₃
MW: 332,015 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07368785
CAS nummer: 886762-67-0