2-[(Trifluoromethyl)thio]benzoic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-[(Trifluoromethyl)sulphanyl]benzoic acid, 2-Carboxyphenyl trifluoromethyl sulphide
PC9435-1G PC9435-5G PC9435-250MG
APOSPC9435-1GEA 550 SEK
APOSPC9435-1G APOSPC9435-5G APOSPC9435-250MG
2-[(Trifluoromethyl)thio]benzoic acid 98%
2-[(Trifluoromethyl)thio]benzoic acid
Formel: C₈H₅F₃O₂S
MW: 222,184 g/mol
Smältpunkt: 120…122 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00236336
CAS nummer: 37526-67-3