2-(4-Fluorobenzenesulphamido)propanoic acid
Leverantör: Apollo Scientific
Varning
Synonyms: N-(4-Fluorobenzenesulphonyl)alanine
PC9378-1G PC9378-250MG
APOSPC9378-1GEA 2250 SEK
APOSPC9378-1G APOSPC9378-250MG
2-(4-Fluorobenzenesulphamido)propanoic acid
2-(4-Fluorobenzenesulphamido)propanoic acid
Formel: C₉H₁₀FNO₄S
MW: 247.243 g/mol
Smältpunkt: 163…165 °C
Storage Temperature: Omgivande
MDL Number: MFCD00664348
CAS nummer: 780-97-2