4,4,5,5,6,6,6-Heptafluoro-1-(thien-2-yl)hexane-1,3-dione
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
1,3-Dioxo-4,4,5,5,6,6,6-heptafluoro-1-(thien-2-yl)hexane, 2-[(Heptafluorobutanoyl)acetyl]thiophene
PC9201-1G
PC9201-250MG
APOSPC9201-1GEA
720
SEK
APOSPC9201-1G
APOSPC9201-250MG
4,4,5,5,6,6,6-Heptafluoro-1-(thien-2-yl)hexane-1,3-dione
4,4,5,5,6,6,6-Heptafluoro-1-(thien-2-yl)hexane-1,3-dione
Formel:
C₁₀H₅F₇O₂S MW: 322,199 g/mol Kokpunkt: 271,5 °C Densitet: 1,566 g/cm³ (25 °C) Storage Temperature: Kylskåp |
MDL Number:
MFCD00047091 CAS nummer: 559-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...