Ethyl 3-fluoro-4-methylbenzoate 95%

Leverantör: Apollo Scientific

PC911426-10G PC911426-1G PC911426-5G
APOSPC911426-10GEA 840 SEK
APOSPC911426-10G APOSPC911426-1G APOSPC911426-5G
Ethyl 3-fluoro-4-methylbenzoate 95%
Ethyl 3-fluoro-4-methylbenzoate
Formel: C₁₀H₁₁FO₂
MW: 182,192 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06204523
CAS nummer: 86239-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR