4-(3-Fluoro-phenyl)-piperidine 95%

Leverantör: Apollo Scientific
Varning

PC911387-1G PC911387-250MG
APOSPC911387-1GEA 6775 SEK
APOSPC911387-1G APOSPC911387-250MG
4-(3-Fluoro-phenyl)-piperidine 95%
4-(3-Fluoro-phenyl)-piperidine
Formel: C₁₁H₁₄FN
MW: 179,234 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05189920
CAS nummer: 104774-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR