3-Cyano-3-(4-fluoro-phenyl)-propionic acid 96%
Leverantör: Apollo Scientific
PC911150-1G PC911150-5G
APOSPC911150-1GEA 2925 SEK
APOSPC911150-1G APOSPC911150-5G
3-Cyano-3-(4-fluoro-phenyl)-propionic acid 96%
3-Cyano-3-(4-fluoro-phenyl)-propionic acid
Formel: C₁₀H₈FNO₂
MW: 193.174 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11036172
CAS nummer: 1267271-26-0