1,2-Dichloro-4-(trifluoromethoxy)benzene 97%
Leverantör: Apollo Scientific
PC910151-1G PC910151-500MG
APOSPC910151-1GEA 5500 SEK
APOSPC910151-1G APOSPC910151-500MG
1,2-Dichloro-4-(trifluoromethoxy)benzene 97%
1,2-Dichloro-4-(trifluoromethoxy)benzene
Formel: C₇H₃Cl₂F₃O
MW: 230.999 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD22199810
CAS nummer: 151276-10-7