1-(4-Fluorophenyl)piperidin-4-one 95%

Leverantör: Apollo Scientific

PC905449-1G PC905449-250MG
APOSPC905449-1GEA 5900 SEK
APOSPC905449-1G APOSPC905449-250MG
1-(4-Fluorophenyl)piperidin-4-one 95%
1-(4-Fluorophenyl)piperidin-4-one
Formel: C₁₁H₁₂FNO
MW: 193,217 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05864502
CAS nummer: 116247-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR