3-Fluorobenzoyl cyanide 95%
Leverantör: Apollo Scientific
PC905425-1G
PC905425-5G
APOSPC905425-1GEA
2675
SEK
APOSPC905425-1G
APOSPC905425-5G
3-Fluorobenzoyl cyanide 95%
3-Fluorobenzoyl cyanide
Formel:
C₈H₄FNO MW: 149,122 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD06658450 CAS nummer: 658-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...