5-Chloro-2-fluoro-3-(hydroxymethyl)pyridine 95%
Leverantör: Apollo Scientific
PC905338-1G PC905338-250MG
APOSPC905338-1GEA 4700 SEK
APOSPC905338-1G APOSPC905338-250MG
5-Chloro-2-fluoro-3-(hydroxymethyl)pyridine 95%
5-Chloro-2-fluoro-3-(hydroxymethyl)pyridine
Formel: C₆H₅ClFNO
MW: 161.561 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04972393
CAS nummer: 884494-79-5