2-Bromo-5-chloro-3-fluoroisonicotinic acid 97%

Leverantör: Apollo Scientific
Varning

PC905149-250MG
APOSPC905149-250MGEA 1175 SEK
APOSPC905149-250MG
2-Bromo-5-chloro-3-fluoroisonicotinic acid 97%
2-Bromo-5-chloro-3-fluoroisonicotinic acid
Formel: C₆H₂BrClFNO₂
MW: 254.441 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD20921503
CAS nummer: 514798-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR