4-Chloro-2,5-difluoroaniline 98%
Leverantör: Apollo Scientific
Synonyms: 4-Chloro-2,5-difluorophenylamine
PC903815-10G PC903815-1G PC903815-5G
APOSPC903815-10GEA 10450 SEK
APOSPC903815-10G APOSPC903815-1G APOSPC903815-5G
4-Chloro-2,5-difluoroaniline 98%
4-Chloro-2,5-difluoroaniline
Formel: C₆H₄ClF₂N
MW: 163,552 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06808807
CAS nummer: 2613-30-1