1,5-Difluoro-2-methyl-4-nitrobenzene 96%
Leverantör: Apollo Scientific
Synonyms: 2,4-Difluoro-5-nitrotoluene
PC902246-1G PC902246-5G
APOSPC902246-1GEA 615 SEK
APOSPC902246-1G APOSPC902246-5G
1,5-Difluoro-2-methyl-4-nitrobenzene 96%
1,5-Difluoro-2-methyl-4-nitrobenzene
Formel: C₇H₅F₂NO₂
MW: 173.117 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16620288
CAS nummer: 179011-38-2