1-Isopropyl-3-(trifluoromethyl)pyrazole 95%
Leverantör: Apollo Scientific
PC902058-1G
PC902058-5G
APOSPC902058-1GEA
2575
SEK
APOSPC902058-1G
APOSPC902058-5G
1-Isopropyl-3-(trifluoromethyl)pyrazole 95%
1-Isopropyl-3-(trifluoromethyl)pyrazole
Formel:
C₇H₉F₃N₂ MW: 178,155 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD16619956 CAS nummer: 915377-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...