Methyl 1-(4-fluorophenyl)cyclopropanecarboxylate 98%
Leverantör: Apollo Scientific
PC901983-10G PC901983-1G PC901983-5G
APOSPC901983-10GEA 8925 SEK
APOSPC901983-10G APOSPC901983-1G APOSPC901983-5G
Methyl 1-(4-fluorophenyl)cyclopropanecarboxylate 98%
Methyl 1-(4-fluorophenyl)cyclopropanecarboxylate
Formel: C₁₁H₁₁FO₂
MW: 194,202 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18434442
CAS nummer: 943111-83-9