5-Fluoro-2-methoxy-3-nitropyridine 96%
Leverantör: Apollo Scientific
PC900607-100MG
PC900607-1G
PC900607-250MG
APOSPC900607-100MGEA
3025
SEK
APOSPC900607-100MG
APOSPC900607-1G
APOSPC900607-250MG
5-Fluoro-2-methoxy-3-nitropyridine 96%
5-Fluoro-2-methoxy-3-nitropyridine
Formel:
C₆H₅FN₂O₃ MW: 172,114 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD18260977 CAS nummer: 1211534-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...