Ethyl 3,3,3-trifluoro-2-(hydroxyimino)propanoate 95%

Leverantör: Apollo Scientific

PC900165-1G PC900165-5G
APOSPC900165-1GEA 2575 SEK
APOSPC900165-1G APOSPC900165-5G
Ethyl 3,3,3-trifluoro-2-(hydroxyimino)propanoate 95%
Ethyl 3,3,3-trifluoro-2-(hydroxyimino)propanoate
Formel: C₅H₆F₃NO₃
MW: 185,101 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD24448807
CAS nummer: 263750-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR