1-Bromo-3-(difluoromethoxy)-5-fluorobenzene 97%
Leverantör: Apollo Scientific
Varning
PC900077-1G PC900077-5G
APOSPC900077-1GEA 1175 SEK
APOSPC900077-1G APOSPC900077-5G
1-Bromo-3-(difluoromethoxy)-5-fluorobenzene 97%
1-Bromo-3-(difluoromethoxy)-5-fluorobenzene
Formel: C₇H₄BrF₃O
MW: 241.005 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD18390899
CAS nummer: 433939-28-7