2-Hydroxy-6-(trifluoromethyl)nicotinonitrile 97%
Leverantör: Apollo Scientific
Synonyms: 3-Cyano-2-hydroxy-6-(trifluoromethyl)pyridine
PC8691-1G PC8691-5G
APOSPC8691-1GEA 2500 SEK
APOSPC8691-1G APOSPC8691-5G
2-Hydroxy-6-(trifluoromethyl)nicotinonitrile 97%
2-Hydroxy-6-(trifluoromethyl)nicotinonitrile
Formel: C₇H₃F₃N₂O
MW: 188,107 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09832863
CAS nummer: 116548-04-0