4-Bromo-3-fluorobenzoyl chloride

Leverantör: Apollo Scientific
Fara

PC8605-1G PC8605-5G
APOSPC8605-1GEA 2075 SEK
APOSPC8605-1G APOSPC8605-5G
4-Bromo-3-fluorobenzoyl chloride
4-Bromo-3-fluorobenzoyl chloride
Formel: C₇H₃BrClFO
MW: 237,454 g/mol
Kokpunkt: 246 °C
Densitet: 1,743 g/cm³ (25 °C)
Flampunkt: 103 °C
Storage Temperature: Omgivande
MDL Number: MFCD09746051
CAS nummer: 695188-21-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR