2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzyl alcohol 97%
Leverantör: Apollo Scientific
Varning
Synonyms: {2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]phenyl}methanol
PC8554-1G PC8554-250MG
APOSPC8554-1GEA 2250 SEK
APOSPC8554-1G APOSPC8554-250MG
2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzyl alcohol 97%
2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzyl alcohol
Formel: C₁₂H₁₁F₃N₂O
MW: 256.224 g/mol
Smältpunkt: 42.5…44 °C
Storage Temperature: Omgivande
MDL Number: MFCD09817511
CAS nummer: 892502-29-3
UN: 2811
ADR: 6.1 III