Perfluorophenyl 3,4-dihydro-4-methyl-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 3,4-Dihydro-4-methyl-7-[(pentafluorophenoxy)carbonyl]-2H-pyrido[3,2-b][1,4]oxazine, Pentafluorophenyl 3,4-dihydro-4-methyl-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylate
PC8541-1G PC8541-250MG
APOSPC8541-1GEA 3775 SEK
APOSPC8541-1G APOSPC8541-250MG
Perfluorophenyl 3,4-dihydro-4-methyl-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylate 97%
Perfluorophenyl 3,4-dihydro-4-methyl-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylate
Formel: C₁₅H₉F₅N₂O₃
MW: 360.236 g/mol
Smältpunkt: 129.5…132 °C
Storage Temperature: Omgivande
MDL Number: MFCD09817508
CAS nummer: 921938-83-2
UN: 2811
ADR: 6.1 III