1H-Nonafluoropentane-1,1-diol
Leverantör: Apollo Scientific
Varning
Synonyms: Nonafluoropentaldehyde hydrate (diol), 2,2,3,3,4,4,5,5,5-Nonafluoropentane-1,1-diol
PC8268-25G PC8268-5G
APOSPC8268-25GEA 6575 SEK
APOSPC8268-25G APOSPC8268-5G
1H-Nonafluoropentane-1,1-diol
1H-Nonafluoropentane-1,1-diol
Formel: C₅H₃F₉O₂
MW: 266,062 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07784233
CAS nummer: 355-30-6