1-[3-(4-Fluorophenoxy)prop-1-yl]piperazine dihydrochloride 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Fluoro-4-[3-(piperazin-1-yl)propoxy]benzene dihydrochloride
PC7936-1G PC7936-2G PC7936-5G PC7936-10G
APOSPC7936-1GEA 0 SEK
APOSPC7936-1G APOSPC7936-2G APOSPC7936-5G APOSPC7936-10G
1-[3-(4-Fluorophenoxy)prop-1-yl]piperazine dihydrochloride 98%
1-[3-(4-Fluorophenoxy)prop-1-yl]piperazine dihydrochloride
Formel: C₁₃H₁₉FN₂O·2ClH
MW: 311,223 g/mol
Smältpunkt: 212…214 °C
Storage Temperature: Omgivande
MDL Number: MFCD03428570
CAS nummer: 537037-78-8