5-Fluoro-DL-tryptophan ethyl ester
Leverantör: Apollo Scientific
Varning
Synonyms: 3-(2-Amino-3-ethoxy-3-oxoprop-1-yl)-5-fluoro-1H-indole, Ethyl 2-amino-3-(5-fluoro-1H-indol-3-yl)propanoate
PC7894-100MG PC7894-50MG
APOSPC7894-100MGEA 1050 SEK
APOSPC7894-100MG APOSPC7894-50MG
5-Fluoro-DL-tryptophan ethyl ester
5-Fluoro-DL-tryptophan ethyl ester
Formel: C₁₃H₁₅FN₂O₂
MW: 250.269 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD09921638
CAS nummer: 154170-01-1