2-(Aminomethyl)-3-(trifluoromethyl)pyridine
Leverantör: Apollo Scientific
Varning
Synonyms: [3-(Trifluoromethyl)pyridin-2-yl]methylamine
PC7173-1G PC7173-250MG PC7173-5G
APOSPC7173-1GEA 2750 SEK
APOSPC7173-1G APOSPC7173-250MG APOSPC7173-5G
2-(Aminomethyl)-3-(trifluoromethyl)pyridine
2-(Aminomethyl)-3-(trifluoromethyl)pyridine
Formel: C₇H₇F₃N₂
MW: 176,139 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07375385
CAS nummer: 886371-24-0