Perfluoro-3,5,6-trichlorohexanoic acid 95%
Leverantör: Apollo Scientific
Fara
Synonyms: 2,2,3,4,4,5,6,6-Octafluoro-3,5,6-trichlorohexanoic acid, 3,5,6-Trichlorooctafluorohexanoic acid
PC6853-25G PC6853-5G
APOSPC6853-25GEA 7725 SEK
APOSPC6853-25G APOSPC6853-5G
Perfluoro-3,5,6-trichlorohexanoic acid 95%
Perfluoro-3,5,6-trichlorohexanoic acid
Formel: C₆HCl₃F₈O₂
MW: 363,417 g/mol
Kokpunkt: 133…135 °C (20 mm Hg)
Densitet: 1,86 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00236081
CAS nummer: 2106-54-9
UN: 3265
ADR: 8,II