Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoic acid 95%
Leverantör: Apollo Scientific
Fara
Synonyms: Perfluoro-2,5-dimethyl-3,6-dioxa-8-propoxynonanoic acid
PC6851-25G PC6851-5G PC6851-1G
APOSPC6851-25GEA 3875 SEK
APOSPC6851-25G APOSPC6851-5G APOSPC6851-1G
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoic acid 95%
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoic acid
Formel: C₁₂HF₂₃O₅
MW: 662,097 g/mol
Kokpunkt: 75…77 °C
Densitet: 1,8 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00236739
CAS nummer: 65294-16-8
UN: 3265
ADR: 8,III