5-Cyano-2-fluorobenzoic acid

Leverantör: Apollo Scientific
Varning

Synonyms: 3-Carboxy-4-fluorobenzonitrile

PC5926-25G PC5926-5G
APOSPC5926-25GEA 1425 SEK
APOSPC5926-25G APOSPC5926-5G
5-Cyano-2-fluorobenzoic acid
5-Cyano-2-fluorobenzoic acid
Formel: C₈H₄FNO₂
MW: 165,121 g/mol
Smältpunkt: 187…189 °C
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD03094321
CAS nummer: 146328-87-2
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR