Pentafluorophenyl isonicotinate 97%

Leverantör: Apollo Scientific
Varning

Synonyms: Pentafluorophenyl pyridine-4-carboxylate

PC5808-1G PC5808-250MG PC5808-5G
APOSPC5808-1GEA 2875 SEK
APOSPC5808-1G APOSPC5808-250MG APOSPC5808-5G
Pentafluorophenyl isonicotinate 97%
Pentafluorophenyl isonicotinate
Formel: C₁₂H₄F₅NO₂
MW: 289,158 g/mol
Smältpunkt: 47,5…55.5 °C
Storage Temperature: Omgivande
MDL Number: MFCD02916455
CAS nummer: 360574-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR