2’-Hydroxy-2,2,2-trifluoroacetophenone 98%
Leverantör: Apollo Scientific
Varning
PC57070-1G PC57070-250MG PC57070-5G
APOSPC57070-1GEA 975 SEK
APOSPC57070-1G APOSPC57070-250MG APOSPC57070-5G
2’-Hydroxy-2,2,2-trifluoroacetophenone 98%
2’-Hydroxy-2,2,2-trifluoroacetophenone
Formel: C₈H₅F₃O₂
MW: 190,119 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06657950
CAS nummer: 25666-51-7