2-(Difluoromethyl)aniline hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Aminobenzal fluoride hydrochloride, α,α-Difluoro-o-toluidine hydrochloride
PC57056-1G PC57056-250MG PC57056-5G
APOSPC57056-1GEA 1975 SEK
APOSPC57056-1G APOSPC57056-250MG APOSPC57056-5G
2-(Difluoromethyl)aniline hydrochloride
2-(Difluoromethyl)aniline hydrochloride
Formel: C₇H₇F₂N·ClH
MW: 179,595 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD23378513
CAS nummer: 1422344-12-4