tert-Butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate
Leverantör: Apollo Scientific
Varning
PC56947-1G PC56947-25G PC56947-5G
APOSPC56947-1GEA 380 SEK
APOSPC56947-1G APOSPC56947-25G APOSPC56947-5G
tert-Butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate
tert-Butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate
Formel: C₁₀H₁₅F₂NO₃·H₂O
MW: 253,243 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD19443979
CAS nummer: 1400264-85-8