6’-Bromo-2,2,2,2’,3’-pentafluoroacetophenone 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-(6-Bromo-2,3-difluorophenyl)-2,2,2-trifluoroethanone
PC56925-1G PC56925-250MG PC56925-5G
APOSPC56925-1GEA 6250 SEK
APOSPC56925-1G APOSPC56925-250MG APOSPC56925-5G
6’-Bromo-2,2,2,2’,3’-pentafluoroacetophenone 97%
6’-Bromo-2,2,2,2’,3’-pentafluoroacetophenone
Formel: C₈H₂BrF₅O
MW: 288,997 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12922590
CAS nummer: 1208076-48-5