1-Bromo-3-nitro-5-(trifluoromethoxy)benzene 95%
Leverantör: Apollo Scientific
Varning
PC56910-1G PC56910-250MG PC56910-5G
APOSPC56910-1GEA 6600 SEK
APOSPC56910-1G APOSPC56910-250MG APOSPC56910-5G
1-Bromo-3-nitro-5-(trifluoromethoxy)benzene 95%
1-Bromo-3-nitro-5-(trifluoromethoxy)benzene
Formel: C₇H₃BrF₃NO₃
MW: 286.003 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28746505
CAS nummer: 1807151-57-0