2,4-Difluoro-3-(trifluoromethoxy)phenylacetylene 95%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Ethynyl-2,4-difluoro-3-(trifluoromethoxy)benzene
PC56733-1G PC56733-250MG
APOSPC56733-1GEA 7700 SEK
APOSPC56733-1G APOSPC56733-250MG
2,4-Difluoro-3-(trifluoromethoxy)phenylacetylene 95%
2,4-Difluoro-3-(trifluoromethoxy)phenylacetylene
Formel: C₉H₃F₅O
MW: 222.112 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD31618862
CAS nummer: 2237235-34-4
UN: 1993
ADR: 3 III