2-(Difluoromethoxy)-5-iodo-3-(trifluoromethyl)pyridine 95%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-(Difluoromethoxy)-5-iodo-3-(trifluoromethyl)pyridine
PC56277-1G PC56277-250MG PC56277-5G
APOSPC56277-1GEA 1100 SEK
APOSPC56277-1G APOSPC56277-250MG APOSPC56277-5G
2-(Difluoromethoxy)-5-iodo-3-(trifluoromethyl)pyridine 95%
2-(Difluoromethoxy)-5-iodo-3-(trifluoromethyl)pyridine
Formel: C₇H₃F₅INO
MW: 339.001 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28789909
CAS nummer: 1803729-60-3