3-Bromo-2-hydroxy-5-(pentafluorothio)benzoic acid 95+%
Leverantör: Apollo Scientific
Fara
Synonyms: 3-Bromo-2-hydroxy-5-(pentafluorosulphanyl)benzoic acid
PC56144-100MG PC56144-250MG
APOSPC56144-100MGEA 1725 SEK
APOSPC56144-100MG APOSPC56144-250MG
3-Bromo-2-hydroxy-5-(pentafluorothio)benzoic acid 95+%
3-Bromo-2-hydroxy-5-(pentafluorothio)benzoic acid
Formel: C₇H₄BrF₅O₃S
MW: 343.066 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD31561290
UN: 2811
ADR: 6.1 III