5-Amino-2-fluoroisonicotinic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Amino-2-fluoropyridine-4-carboxylic acid
PC5496-1G PC5496-250MG
APOSPC5496-1GEA 2025 SEK
APOSPC5496-1G APOSPC5496-250MG
5-Amino-2-fluoroisonicotinic acid
5-Amino-2-fluoroisonicotinic acid
Formel: C₆H₅FN₂O₂
MW: 156,115 g/mol
Smältpunkt: 259 °C
Storage Temperature: Omgivande
MDL Number: MFCD03411724
CAS nummer: 171178-43-1