Methyl 4,5-difluoro-2-nitro-benzoate 95%
Leverantör: Apollo Scientific
Synonyms: Methyl 4,5-difluoro-2-nitrobenzoate
PC53622-25G PC53622-5G
APOSPC53622-25GEA 2250 SEK
APOSPC53622-25G APOSPC53622-5G
Methyl 4,5-difluoro-2-nitro-benzoate 95%
Methyl 4,5-difluoro-2-nitro-benzoate
Formel: C₈H₅F₂NO₄
MW: 217,126 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11042825
CAS nummer: 1015433-96-1