3-Bromo-2,6-difluorobenzal fluoride
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Bromo-3-(difluoromethyl)-2,4-difluorobenzene
PC53560-1G PC53560-250MG PC53560-5G
APOSPC53560-1GEA 1250 SEK
APOSPC53560-1G APOSPC53560-250MG APOSPC53560-5G
3-Bromo-2,6-difluorobenzal fluoride
3-Bromo-2,6-difluorobenzal fluoride
Formel: C₇H₃BrF₄
MW: 242.996 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD25976517
CAS nummer: 1447671-68-2