2,6-Dichloro-4-fluorobenzonitrile 98%
Leverantör: Apollo Scientific
Varning
PC53493-1G PC53493-5G PC53493-25G
APOSPC53493-1GEA 295 SEK
APOSPC53493-1G APOSPC53493-5G APOSPC53493-25G
2,6-Dichloro-4-fluorobenzonitrile 98%
2,6-Dichloro-4-fluorobenzonitrile
Formel: C₇H₂Cl₂FN
MW: 190,002 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28064264
CAS nummer: 1473423-59-4