2,6-Difluoro-3-(trifluoromethyl)benzyl bromide
Leverantör: Apollo Scientific
Fara
Synonyms: 2-(Bromomethyl)-1,3-difluoro-4-(trifluoromethyl)benzene
PC53481-1G PC53481-250MG PC53481-5G
APOSPC53481-1GEA 3875 SEK
APOSPC53481-1G APOSPC53481-250MG APOSPC53481-5G
2,6-Difluoro-3-(trifluoromethyl)benzyl bromide
2,6-Difluoro-3-(trifluoromethyl)benzyl bromide
Formel: C₈H₄BrF₅
MW: 275.013 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD30723303
CAS nummer: 2149589-86-4
UN: 3265
ADR: 8 III