Methyl 3-bromo-4,5-difluorobenzoate
Leverantör: Apollo Scientific
Varning
PC53467-1G PC53467-5G PC53467-10G
APOSPC53467-1GEA 360 SEK
APOSPC53467-1G APOSPC53467-5G APOSPC53467-10G
Methyl 3-bromo-4,5-difluorobenzoate
Methyl 3-bromo-4,5-difluorobenzoate
Formel: C₈H₅BrF₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD20483155
CAS nummer: 1244642-70-3