3-(Trifluoromethyl)benzoyl isothiocyanate
Leverantör: Apollo Scientific
PC53358-1G PC53358-250MG
APOSPC53358-1GEA 6825 SEK
APOSPC53358-1G APOSPC53358-250MG
3-(Trifluoromethyl)benzoyl isothiocyanate
3-(Trifluoromethyl)benzoyl isothiocyanate
Formel: C₉H₄F₃NOS
MW: 231,194 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12188974
CAS nummer: 100663-25-0