3-Bromo-4,5-difluorobenzoic acid 99%
Leverantör: Apollo Scientific
Varning
PC53246-1G PC53246-25G PC53246-5G
APOSPC53246-1GEA 380 SEK
APOSPC53246-1G APOSPC53246-25G APOSPC53246-5G
3-Bromo-4,5-difluorobenzoic acid 99%
3-Bromo-4,5-difluorobenzoic acid
Formel: C₇H₃BrF₂O₂
MW: 236,998 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD20233291
CAS nummer: 1244642-73-6