Methyl 3-nitro-4-(trifluoromethoxy)benzoate
Leverantör: Apollo Scientific
Varning
PC53155-1G PC53155-250MG PC53155-5G
APOSPC53155-1GEA 3300 SEK
APOSPC53155-1G APOSPC53155-250MG APOSPC53155-5G
Methyl 3-nitro-4-(trifluoromethoxy)benzoate
Methyl 3-nitro-4-(trifluoromethoxy)benzoate
Formel: C₉H₆F₃NO₅
Storage Temperature: Omgivande
MDL Number: MFCD14698078
CAS nummer: 728-54-1