3-Bromo-2,6-difluoro-5-(trifluoromethyl)benzaldehyde, tech
Leverantör: Apollo Scientific
|
Varning
|
PC53111-1G
PC53111-250MG
APOSPC53111-1GEA
8875
SEK
APOSPC53111-1G
APOSPC53111-250MG
3-Bromo-2,6-difluoro-5-(trifluoromethyl)benzaldehyde, tech
3-Bromo-2,6-difluoro-5-(trifluoromethyl)benzaldehyde
Formel:
C₈H₂BrF₅O MW: 288.997 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD29036692 CAS nummer: 1980053-04-0 |