(R)-2-Amino-2-(4-trifluoromethylphenyl)ethanol 97%
Leverantör: Apollo Scientific
Synonyms: (R)-2-Amino-2-(4-(trifluoromethyl)phenyl)ethanol
PC530039-1G PC530039-250MG PC530039-500MG
APOSPC530039-1GEA 12850 SEK
APOSPC530039-1G APOSPC530039-250MG APOSPC530039-500MG
(R)-2-Amino-2-(4-trifluoromethylphenyl)ethanol 97%
(R)-2-Amino-2-(4-trifluoromethylphenyl)ethanol
Formel: C₉H₁₀F₃NO
MW: 205,177 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09253775
CAS nummer: 306281-86-7